Thermo Scientific Chemicals

4-Bromophenyl chloroformate, 95+%, Thermo Scientific Chemicals

Catalog number: 430150050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Bromophenyl chloroformate, 95+%, Thermo Scientific Chemicals

Catalog number: 430150050
5 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
7693-44-9
IUPAC Name
4-bromophenyl carbonochloridate
Molecular Formula
C7H4BrClO2
InChI Key
IKMNJYGTSSQNSE-UHFFFAOYSA-N
SMILES
ClC(=O)OC1=CC=C(Br)C=C1
Appearance (Form)
Low melting solid or liquid
Infrared spectrum
Conforms
GC
>=95.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text