1-Cyclohexenylboronic acid pinacol ester, 97%
1-Cyclohexenylboronic acid pinacol ester, 97%
1-Cyclohexenylboronic acid pinacol ester, 97%
Thermo Scientific Chemicals

1-Cyclohexenylboronic acid pinacol ester, 97%

CAS: 141091-37-4 | C12H21BO2 | 208.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
Catalog number 430315000
Price (USD)
150.00
Each
Quantity:
500 mg
Request bulk or custom format
Price (USD)
150.00
Each
Chemical Identifiers
CAS141091-37-4
IUPAC Name2-(cyclohex-1-en-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular FormulaC12H21BO2
InChI KeyQNZFUMVTUFOLRT-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to tan
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only