Product Image
Applied Biosystems™

Glass Syringe, 1.0 mL

This Glass Syringe is suitable for filling 310 Genetic Analyzer capillaries with POP-4™ Performance Optimized Polymer, POP-6™ Performance Optimized Polymer,Read more
Have Questions?
Catalog NumberQuantity
43044711 syringe
Catalog number 4304471
Price (USD)
195.00
Each
Quantity:
1 syringe
Price (USD)
195.00
Each
This Glass Syringe is suitable for filling 310 Genetic Analyzer capillaries with POP-4™ Performance Optimized Polymer, POP-6™ Performance Optimized Polymer, and GeneScan™ polymer.

Used With
310 Genetic Analyzer Capillary, 47 cm (36 cm wtr), 310 Running Buffer, 10X, BigDye™ Terminator v1.1 Cycle Sequencing Kit, BigDye™ Terminator v3.1 Cycle Sequencing Kit, Genetic Analyzer Sample Tubes, 0.5 ml, Hi-Di™ Formamide, POP-4™ Polymer for 3100/3100-Avant™ Genetic Analyzers, POP-4™ Polymer for the 310 Genetic Analyzer, POP-6™ Polymer for the 310 Genetic Analyzer, Septa for 0.5 ml Sample Tubes.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeSyringe
Quantity1 syringe
For Use With (Equipment)310 GeneticAnalyzer
MaterialGlass
Volume (Metric)1 mL
Unit SizeEach

Frequently asked questions (FAQs)

How often should I change my polymer and buffer? 

Polymer in the syringe should be changed every three days. Do not return unused polymer to the bottle.

Buffer and water should be changed every 48 hours or, if the run lasts for more than 48 hours, after the run is complete.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.

How often do I really need to clean the pump block?

The pump block needs to be cleaned when:

Installing a syringe.

Switching applications (i.e., from sequencing to fragment analysis).

Shutting down the instrument for a long period of time.

If the instrument has run for more than 4 days without any of the above occurring.

Appearance of unusual spikes in the electropherogram.

If the syringe containing the polymer has been on the instrument for more than a week, it is important to clean the syringe and the block, and to change the polymer. At room temperature, enough urea decomposition can occur in one week to cause transient current increases during electrophoreses. If polymer is allowed to dry in the pump block channel, the pump block may be ruined.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.

What happens to the 310 Genetic Analyzer if it runs out of polymer during the run?

A message will appear at the start of the run telling you if you have enough polymer to complete the run. If you should continue without adding more polymer, then the run will be cancelled at the injection where it ran out and an entry will be made into the run log that there is no polymer. The following injections will appear to run for 1–2 minutes each and will be recorded into the run log. No data will be collected for these injections.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.