2-Isopropenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 90%
2-Isopropenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 90%
2-Isopropenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 90%
Thermo Scientific Chemicals

2-Isopropenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 90%

CAS: 126726-62-3 | C9H17BO2 | 168.04 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4304500505 gGlass bottle
Catalog number 430450050
Price (USD)
121.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
121.00
Each
Chemical Identifiers
CAS126726-62-3
IUPAC Name4,4,5,5-tetramethyl-2-(prop-1-en-2-yl)-1,3,2-dioxaborolane
Molecular FormulaC9H17BO2
InChI KeySVSUYEJKNSMKKW-UHFFFAOYSA-N
SMILESCC(=C)B1OC(C)(C)C(C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=88.0 %
Appearance (Color)Clear colorless to brown
Infrared spectrumConforms
StabilizerPhenothiazine stabilizer
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only