1-Methylindazole-6-boronic acid, 97%
1-Methylindazole-6-boronic acid, 97%
1-Methylindazole-6-boronic acid, 97%
Thermo Scientific Chemicals

1-Methylindazole-6-boronic acid, 97%

CAS: 1150114-80-9 | C8H9BN2O2 | 175.98 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4304800101 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 430480010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1150114-80-9
IUPAC Name(1-methyl-1H-indazol-6-yl)boronic acid
Molecular FormulaC8H9BN2O2
InChI KeyOMVQAWCHQBKZCF-UHFFFAOYSA-N
SMILESCN1N=CC2=CC=C(C=C12)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only