Ácido (5-trifluorometilpiridin-3-il)borónico, 95 %, Thermo Scientific Chemicals
Ácido (5-trifluorometilpiridin-3-il)borónico, 95 %, Thermo Scientific Chemicals
Ácido (5-trifluorometilpiridin-3-il)borónico, 95 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ácido (5-trifluorometilpiridin-3-il)borónico, 95 %, Thermo Scientific Chemicals

CAS: 947533-51-9 | C6H5BF3NO2 | 190.92 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
250 mg
1 g
Envase:
Frasco de vidrio
Número de catálogo 430522500
Precio (EUR)
179,00
Each
Cantidad:
250 mg
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Precio (EUR)
179,00
Each
Identificadores químicos
CAS947533-51-9
IUPAC Name[5-(trifluoromethyl)pyridin-3-yl]boronic acid
Molecular FormulaC6H5BF3NO2
InChI KeySFBQNNGMEKUJAN-UHFFFAOYSA-N
SMILESOB(O)C1=CC(=CN=C1)C(F)(F)F
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)White to yellow
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
HPLC>=94.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only