AbsoluteRNA Wash Solution

Catalog number: 4305545

Invitrogen™  Related applications: RNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This is an on-column, in protocol DNase treatment reagent for total RNA.

Total RNA Chemistry allows rapid isolation of total RNA from various samples, including tissue-cultured cells, whole blood, and PBMC. The protocol is designed for use with the 6100 Nucleic Acid PrepStation and 6700 Automated Nucleic Acid Workstation.

Total RNA Chemistry uses a unique formulation to effectively lyse cells. Lysis occurs almost immediately after cultured cells or homogenized tissues are mixed with lysis buffer. The reagent inactivates cellular RNases, preventing degradation of the isolated nucleic acid while keeping gDNA in the solution. This allows a separation of total RNA from gDNA and other cellular debris. The cellular lysate or tissue homogenate is then transferred to the total RNA purification tray, where it is passed through the tray under precisely controlled vacuum conditions.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Buffer Type: Digestion Buffer
Form: Liquid
Format: Tube(s)
Product Line: Tempus™
Product Size: 10 mL
Shipping Condition: Dry Ice

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols