Applied Biosystems™

Septa for 0.2 mL MicroAmp™ tubes (for the 310 Genetic Analyzer)

Catalog number:  4305547

 Related applications:

Sanger Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

These are septa covers for 0.2 ml MicroAmp™ sample tubes, for use with the 310 Genetic Analyzer.

Use with: MicroAmp® 8-Tube Strips (0.2 ml).

For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 310 Genetic Analyzer
Product Line: MicroAmp™
Product Size: 480 each
Shipping Condition: Room Temperature
Tube Type: Assay Tube, Conical Tube, Disposable Tube
Type: Septa

Contents & storage

Each package contains 480 septa.

Documents

Manuals & protocols