Product Image
Applied Biosystems™

BigDye™ Terminator v1.0 & v3.0 5X Sequencing Buffer

This sequencing buffer is optimized for use with BigDye™ Terminator v1.0 or v3.0 cycle sequencing kits.For Research Use Only. NotRead more
Have Questions?
Catalog NumberQuantity
430560310.8 mL
Catalog number 4305603
Price (EUR)
3.699,20
Online exclusive
5.440,00
Save 1.740,80
Each
Quantity:
10.8 mL
Price (EUR)
3.699,20
Online exclusive
5.440,00
Save 1.740,80
Each
This sequencing buffer is optimized for use with BigDye™ Terminator v1.0 or v3.0 cycle sequencing kits.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferSequencing Buffer
Concentration5 X
For Use With (Application)Standard Sequencing
For Use With (Equipment)GeneAmp 9700, Veriti Thermal Cycler, 3500 Genetic Analyzer, 3730xl DNA Analyzer, 3130xl Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer
FormLiquid
Product LineBigDye Terminator, BigDye
Product TypeSequencing Buffer
Quantity10.8 mL
Shipping ConditionDry Ice
Unit SizeEach
Contents & Storage
Store at -15 to -25°C

Frequently asked questions (FAQs)

Are the BigDye Terminator 5X buffer v1.1 & v3.1 and BigDye Terminator 5X buffer v1.0 & v3.0 interchangeable?

The composition of the BigDye Terminator 5X buffer v1.1 & v3.1 and BigDye Terminator 5X buffer v1.0 & v3.0 chemistries is different and therefore should only be used with the matching chemistry. Use of the incorrect BigDye Terminator 5X buffer can negatively impact sequencing results.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.