Product Image
Applied Biosystems™

dGTP BigDye™ Terminator v1.0 & v3.0 5X Sequencing Buffer

This sequencing buffer is optimized for use with BigDye™ Terminator v1.0 or v3.0 cycle sequencing kits.For Research Use Only. NotRead more
Have Questions?
Catalog NumberQuantity
43056051.2 mL
Catalog number 4305605
Price (USD)
604.00
Each
Quantity:
1.2 mL
Price (USD)
604.00
Each
This sequencing buffer is optimized for use with BigDye™ Terminator v1.0 or v3.0 cycle sequencing kits.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferSequencing Buffer
Concentration5 X
For Use With (Application)GT-Rich or Difficult Template Sequencing
For Use With (Equipment)GeneAmp 9700, Veriti Thermal Cycler, 3500 Genetic Analyzer, 3730xl DNA Analyzer, 3130xl Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer
FormLiquid
Product LineBigDye Terminator, BigDye
Product TypeSequencing Buffer
Quantity1.2 mL
Shipping ConditionDry Ice
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).