GeneAmp&trade; 10X PCR Gold Buffer &amp; MgCl<sub>2</sub>
Applied Biosystems™

GeneAmp™ 10X PCR Gold Buffer & MgCl2

Applied Biosystems GeneAmp 10X PCR Gold Buffer is formulated to provide flexible, efficient activation of AmpliTaq Gold DNA Polymerase, resulting in highly specific, robust PCR amplification.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoVolume
43068986 x 1.5 mL
43798742 x 75 mL
Número do catálogo 4306898
Preço (BRL)
-
Volume:
6 x 1.5 mL
Request bulk or custom format
Applied Biosystems GeneAmp 10X PCR Gold Buffer and MgCl2 is formulated to provide flexible, efficient activation of AmpliTaq Gold DNA Polymerase, resulting in highly specific, robust PCR amplification. The ionic strength and pH of GeneAmp 10X PCR Gold Buffer has been optimized to provide a wider activation temperature and time range when used in with AmpliTaq Gold DNA Polymerase.

When GeneAmp 10X PCR Gold Buffer and the MgCl2 solution are used together, there is greater versatility in optimizing magnesium ion concentrations for optimal PCR amplification with any specific set of primers and DNA template.

Features

  • Magnesium ion–free
  • Gelatin-free
  • High versatility in optimizing magnesium ion concentration
  • MgCl2 solution provided separately
For Research Use Only. Not for use in diagnostic procedures.
Especificações
FormatKit
PolymeraseAmpliTaq Gold DNA Polymerase
Product Type10X PCR Gold Buffer
Quantity6 x 1.5 mL
Shipping ConditionDry Ice
Volume6 x 1.5 mL
Concentration10X
For Use With (Application)Standard PCR
Unit SizeEach
Conteúdo e armazenamento
• GeneAmp 10X PCR Gold Buffer, 6 x 1.5 mL
• 25 mM MgCl2, 6 x 1.5 mL

Store at -20°C.