dGTP BigDye™ Terminator Cycle Sequencing Kit with the AmpliTaq™ DNA Polymerase, FS. This Ready Reaction Kit uses the same dyeRead more
Have Questions?
Catalog Number
Quantity
4307175
100 Reactions
Catalog number 4307175
Price (USD)
1,908.00
Each
Quantity:
100 Reactions
Price (USD)
1,908.00
Each
dGTP BigDye™ Terminator Cycle Sequencing Kit with the AmpliTaq™ DNA Polymerase, FS. This Ready Reaction Kit uses the same dye set as the BigDye™ v1.0 kits.
The dGTP BigDye™ Terminator Cycle Sequencing Kit's fluorescent dideoxy terminator chemistry is optimized for sequencing through GT- and G-rich templates.
• Ensure reliable results with difficult regions and finishing applications. • Sequence templates with challenging sequence contexts. • Sequence problematic templates that standard chemistries cannot.
Sequence Challenging Templates The dGTP BigDye™ Terminator Kit is ideal for working with GT- and G-rich and other difficult-to-sequence templates. The dGTP BigDye™ Terminator Kit replaces dITP with dGTP and it enables you to extend through those difficult-to-sequence regions with confidence, avoiding early signal loss in these samples.
Optimized for GT-Rich Templates Obtain higher success rate even with your difficult GT-rich templates. By using the same chemistries as the standard BigDye™ Terminator Kits (only replacing dITP with dGTP) the dGTP BigDye™ Terminator Kit provides proven performance, optimized for GT-rich and other difficult-to-sequence templates.
Easily Integrate into Your Workflow dGTP BigDye™ Terminator Kits require no new software or instrument recalibration, so you can easily integrate it into your current workflow with minimal changes to your protocols.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)GT-Rich or Difficult Template Sequencing
No. of Reactions100 reactions
Product LineBigDye Terminator, BigDye
Product TypeSequencing Kit
Quantity100 Reactions
TechniqueFluorescent Dye Terminator Sequencing
Template CompatibilityBAC DNA, Genomic DNA (Bacterial), Plasmid DNA (≤15Kb), PCR Amplicons, Single Stranded DNA
Format12-well Strips, 8-well Strip, 96- or 384-well Plate
Unit SizeEach
Contents & Storage
The kit contains the AmpliTaq™ DNA Polymerase, FS, BigDye terminators v1.0, pGEM™ - 3Zf(+) Control Template, -21 M13 Control Primer (forward), and Premixed Ready Reaction. Store kit at -15 to -25°C.
Frequently asked questions (FAQs)
What information should I include when submitting data to Genetic Analysis Technical Support?
When contacting Genetic Analysis Support (FAS, FSE, Technical Support), please have (if possible):
Instrument model and serial numbers
Version of the software are you using
Application and/or kit information
Kit lot number
Number of runs on the capillary or array
Polymer type and lot number
Buffer lot number
Hi-Di Formamide lot number or loading solution information
Sample data that can be sent to the support person
Details on:
-Reaction setup (i.e., how much DNA was used, primer, etc.)
-How the reactions were cleaned up (alcohol precipitation, columns, etc)
-What template DNA was used (i.e. plasmid, PCR product)
-How the template DNA was prepared?
All of this information will help the support person quickly and accurately assess the problem and provide you with recommendations for resolution.
I have a long homopolymer stretch in my sample that I cannot get through. Do you have any tips for getting through it?
In order to get through homopolymeric regions, you can either use anchored sequencing primers or try using the dRhodamine Terminator Cycle Sequencing Kit since it uses ddTTP instead of ddUTP and has been shown to be less prone to producing stutters, specifically with poly-T regions.
For more information on sequencing through homopolymer stretches, please refer to the DNA Sequencing by Capillary Electrophoresis: Applied Biosystems Chemistry Guide: Second Edition (Cat. No. 4305080, Rev. C). The guide can be found by searching the Thermo Fisher Scientific website with the catalog number 4305080.
I am trying to sequence GC-rich DNA without success. Do you have any tips for getting through this?
When trying to sequence through GC-rich regions, the following tips may improve your success:
• Set the reaction up at 1X or 0.5X strength. Heavily diluting the BigDye Terminator Ready Reaction mix can reduce the success of the sequecing reaction through long GC stretches.
• Perform a hot start at 98–99°C for 5 minutes.
• Use 5% (w/v) DMSO or freshly made betaine in the reaction.
• Use the dGTP BigDye Terminator Cycle Sequencing Kit. G peaks will appear compressed due to the presence of ddGTP, but sequencing through long GC stretches using the dGTP kit is typically more successful than with the standard BigDye Terminator Cycle Sequencing Kits (containing ddITP).
For more information on sequencing GC-rich DNA, please refer to the DNA Sequencing by Capillary Electrophoresis: Applied Biosystems Chemistry Guide: Second Edition (Cat. No. 4305080, Rev. C). The guide can be found by searching the Thermo Fisher Scientific website with the catalog number 4305080.
When should I use the dGTP BigDye Terminator Cycle Sequencing Kit?
The dGTP BigDye Terminator Cycle Sequencing Kit should be used for difficult templates, such as templates that are GT-rich, G-rich templates or are difficult to sequence with other kits. They should be used only when you cannot obtain good data using standard BigDye Terminator kits.