Product Image
Applied Biosystems™

Total RNA Control (Human)

Human Total RNA is provided at a convenient reverse-transcription–ready concentration for all RT-PCR reactions. TaqMan® Control Total RNA is from자세히 알아보기
Have Questions?
카탈로그 번호수량
4307281100 μL
카탈로그 번호 4307281
제품 가격(KRW)
128,000
온라인 행사
Ends: 31-Dec-2026
150,000
할인액 22,000 (15%)
Each
수량:
100 μL
제품 가격(KRW)
128,000
온라인 행사
Ends: 31-Dec-2026
150,000
할인액 22,000 (15%)
Each
Human Total RNA is provided at a convenient reverse-transcription–ready concentration for all RT-PCR reactions. TaqMan® Control Total RNA is from Raji cells (Burkitt's lymphoma), and is conveniently packaged in a PCR-ready concentration for use as a control template in RT-PCR reactions. The Human total RNA is provided at a concentration of 50 ng/μl.
For Research Use Only. Not for use in diagnostic procedures.
사양
형식Frozen
라벨 또는 염료Unlabeled
수량100 μL
배송 조건Dry Ice
Human
농도50 ng/μL
Unit SizeEach
구성 및 보관
100 μl Total Human RNA (at 50 ng/μl). Store at -15 to -25°C.

자주 묻는 질문(FAQ)

In which buffer is the Total RNA Control (Human) (Cat. No. 4307281) suspended?

The the Total RNA Control (Human) (Cat. No. 4307281) is suspended in 1 X TE buffer.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.