Product Image
Applied Biosystems™

Single-Channel Dual 384-well Temperature Verification Probe for PCR 9700 System, 0.02 mL

Have Questions?
Catalog NumberQuantity
43087571 probe
Catalog number 4308757
Price (USD)
3,025.00
Each
Quantity:
1 probe
Price (USD)
3,025.00
Each
The Temperature Verification Kits and Probes are engineered and manufactured specifically for Applied Biosystems to validate temperature calibration and uniformity in GeneAmp™ PCR and the Veriti™ Thermal Cycler instrument systems. This product is the spare probe for the 0.02mL Single Channel 384-well Temperature Verification Kit (PN 4308354).

• Self-contained digital thermometer and battery operated for ease of use
• Interchangeable Platinum Resistant probes

Complete Offering
To expand the use of an existing Temperature Verification Kits, the individual platinum resistant probes are now available in 0.02 mL, 0.1mL, 0.2 mL, for purchase.

More information can also be found at Alpha Technics (Formerly Alpha Sensors, Inc., Eutechnics Division) (www.alphatechnics.com). Calibration standards are traceable to NIST (US National Institute of Standards and Technology). The kit contains one platinum resistant probe, hand held thermometer, mineral oil, cotton swabs and carrying case.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Real Time PCR (qPCR)
For Use With (Equipment)GeneAmp 9700
FormatKit
Quantity1 probe
Shipping ConditionRoom Temperature
Product TypeTemperature Verification Probe
Unit SizeEach
Contents & Storage
Store at Room Temperature