Dichlorodimethylsilane, 99+%, AcroSeal™
Dichlorodimethylsilane, 99+%, AcroSeal™
Dichlorodimethylsilane, 99+%, AcroSeal™
Thermo Scientific Chemicals

Dichlorodimethylsilane, 99+%, AcroSeal™

CAS: 75-78-5 | C2H6Cl2Si | 129.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 430888000
Price (USD)
77.65
Special offer
Online exclusive
Ends: 28-Dec-2026
90.80
Save 13.15
Each
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
77.65
Special offer
Online exclusive
Ends: 28-Dec-2026
90.80
Save 13.15
Each
Chemical Identifiers
CAS75-78-5
IUPAC Namedichlorodimethylsilane
Molecular FormulaC2H6Cl2Si
InChI KeyLIKFHECYJZWXFJ-UHFFFAOYSA-N
SMILESC[Si](C)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=99.0 %
Appearance (Form)Liquid
Refractive index1.4030 to 1.4060 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only