3-Aminopropyltriethoxysilane, 99%, AcroSeal™
3-Aminopropyltriethoxysilane, 99%, AcroSeal™
3-Aminopropyltriethoxysilane, 99%, AcroSeal™
Thermo Scientific Chemicals

3-Aminopropyltriethoxysilane, 99%, AcroSeal™

CAS: 919-30-2 | C9H23NO3Si | 221.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 430948000
Price (EUR)
313,00
Each
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (EUR)
313,00
Each
Specifications
Assay Percent Range98.5% min. (GC)
CAS Max %100.0
CAS Min %98.5
CAS919-30-2
Chemical Name or Material3-Aminopropyltriethoxysilane
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only