Allyl glycidyl ether, 99+%, AcroSeal™
Allyl glycidyl ether, 99+%, AcroSeal™
Allyl glycidyl ether, 99+%, AcroSeal™
Thermo Scientific Chemicals

Allyl glycidyl ether, 99+%, AcroSeal™

CAS: 106-92-3 | C6H10O2 | 114.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 430958000
Price (EUR)
130,00
Each
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (EUR)
130,00
Each
Specifications
Assay Percent Range99% min. (GC)
CAS Max %100.0
CAS Min %99.0
CAS106-92-3
Chemical Name or MaterialAllyl glycidyl ether
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only