ProPac™ 3R SCX 3 μm HPLC Columns
Thermo Scientific™

ProPac™ 3R SCX 3 μm HPLC Columns

Get the performance you need with ProPac™ 3R SCX HPLC columns. Suitable for charge variant separations and recommended for therapeutic proteins, mAbs, ADCs, and biosimilar proteins.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호길이(미터법)직경(미터법)유속
43103-104068100 mm4 mm0.3 to 0.5 mL/min.
43103-05206850 mm2 mm0.1 to 0.3 mL/min
43103-05406850 mm4 mm0.3 to 0.5 mL/min
43103-102068100 mm2 mm0.1 to 0.2 mL/min.
카탈로그 번호 43103-104068
제품 가격(KRW)
-
길이(미터법):
100 mm
직경(미터법):
4 mm
유속:
0.3 to 0.5 mL/min.

Use Thermo Scientific™ ProPac™ 3R SCX 3 μm HPLC columns to provide the reproducibility, resolution, and robustness needed for protein characterization. These columns are ideal for charge variant analysis and recommended for use with therapeutic proteins, monoclonal antibodies (mAbs), antibody drug conjugates (ADCs), and biosimilar proteins.

Designed with bio-inert materials to reduce secondary interactions, our ProPac 3R SCX columns deliver the performance you need over a broad range of pH's, temperatures, and mobile phase compositions with excellent recovery and low carryover.

Suitable for use with mass spectrometry (MS).

Maximizing Reproducibility, Resolution, and Robustness

The ProPac 3R SCX 3 μm HPLC column is a strong cation-exchanger with a sulfonic acid functional group, suitable for use in biomolecule applications. These specialized ion-exchange chromatography columns provide qualitative and quantitative evaluation of charge variant analysis to biopharmaceutical scientists who need reproducible resolving capabilities when identifying and quantifying charge variant profiles of proteins.

Our ProPac 3R SCX columns offer superior performance for characterization of proteins including monoclonal antibodies (mAbs), therapeutic proteins, antibody drug conjugates (ADCs), and biosimilar proteins.

Column Features:

  • Ideal for characterization and quality control assessment of closely-related protein variants
  • Unequaled high-resolution separations
  • High-efficiency peaks with excellent recovery
  • Higher capacity than other columns based on nonporous particles
  • Superior column-to-column and lot-to-lot reproducibility eliminates column variability as a concern in stability and QA/QC testing
  • Compatible with mass spectrometry (MS)

We recommend our CX-1 pH gradient buffers when using these columns with LC-UV/LC-FLD.

사양
컬럼 형식Analytical Column
컬럼 유형Ion Exchange
유속0.3 to 0.5 mL/min.
최대 압력4500 psi (310 bar)
이동상 호환성Recommended for use with CX-1 pH buffers
팩킹 물질Monodisperse Polymer Resin
정지상Proprietary
온도Ambient to 60°C
직경(미터법)4 mm
길이(미터법)100 mm
물질PEEK Hardware
입자 크기3 μm
Strong Cation Exchange
기공 크기Nonporous
제품라인ProPac
기술Bio-LC
pH2 to 12
Unit SizeEach