1H-Indazole-4-carboxaldehyde, 97%
1H-Indazole-4-carboxaldehyde, 97%
1H-Indazole-4-carboxaldehyde, 97%
Thermo Scientific Chemicals

1H-Indazole-4-carboxaldehyde, 97%

CAS: 669050-70-8 | C8H6N2O | 146.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4310300101 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 431030010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS669050-70-8
IUPAC Name1H-indazole-4-carbaldehyde
Molecular FormulaC8H6N2O
InChI KeyFPJXNCKSPFGQGC-UHFFFAOYSA-N
SMILESO=CC1=C2C=NNC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point165°C to 169°C
Appearance (Form)Crystalline powder
Appearance (Color)White
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only