1,3-Benzothiazole-6-carboxaldehyde, 97%
1,3-Benzothiazole-6-carboxaldehyde, 97%
1,3-Benzothiazole-6-carboxaldehyde, 97%
Thermo Scientific Chemicals

1,3-Benzothiazole-6-carboxaldehyde, 97%

CAS: 19989-67-4 | C8H5NOS | 163.2 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
產品號碼 431150010
價格 (TWD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS19989-67-4
IUPAC Name1,3-benzothiazole-6-carbaldehyde
Molecular FormulaC8H5NOS
InChI KeyAVSFPLJXSHRMHM-UHFFFAOYSA-N
SMILESO=CC1=CC=C2N=CSC2=C1
檢視更多
規格Specification Sheet規格表
Appearance (Color)White to yellow
HPLC>=96.0 %
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
Melting point86°C to 96°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only