kappa-Carrageenan
kappa-Carrageenan
kappa-Carrageenan
Thermo Scientific Chemicals

kappa-Carrageenan

CAS: 11114-20-8 | C24H36O25S2-2 | 788.647 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 431590250
Price (USD)
51.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
51.00
Each
Chemical Identifiers
CAS11114-20-8
IUPAC Name(2R,3R,4R,5R,6S)-6-{[(1R,3R,4R,5R,8S)-3-{[(2S,3R,4R,5S,6R)-2-{[(1R,3S,4R,5R,8S)-3,4-dihydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-3-hydroxy-6-(hydroxymethyl)-5-(sulfonatooxy)oxan-4-yl]oxy}-4-hydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl sulfate
Molecular FormulaC24H36O25S2
InChI KeyZNOZWUKQPJXOIG-XSBHQQIPSA-L
SMILESOC[C@H]1O[C@@H](O[C@H]2[C@H]3CO[C@@H]2[C@@H](O)[C@@H](O[C@@H]2[C@@H](O)[C@H](O[C@H]4[C@H]5CO[C@@H]4[C@@H](O)[C@@H](O)O5)O[C@H](CO)[C@@H]2OS([O-])(=O)=O)O3)[C@H](O)[C@@H](O)[C@H]1OS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Ash=<40 %
Infrared spectrumConforms
Appearance (Color)White to off-white to yellow to brown
Appearance (Form)Powder
Loss on drying=<12 % (1 g, 105°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only