Reservoir Septa

Catalog number: 4315932

Applied Biosystems™  Related applications: Sanger Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This is a Reservoir Septa for ABI PRISM® 3100 Genetic Analyzer and ABI PRISM® 3100-Avant™ Genetic Analyzer.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 3100 Genetic Analyzer, 3100-Avant™ Genetic Analyzer
Product Line: Applied Biosystems™
Product Size: 20 each
Shipping Condition: Room Temperature
Type: Septa

Contents & storage

Each package contains 20 septa. Store at room temperature.

Documents

Manuals & protocols