Applied Biosystems™

DS-30 GeneScan™ Installation Standard

Catalog number:  4316144

Related applications:

Genotyping & Genomic Profiling

| Sanger Sequencing

Back to top

Description

The GeneScan™ Installation Standard DS-30 contains pooled PCR products amplified from microsatellite loci present in CEPH individual 1347. The products are labeled with 6-FAM™, HEX™ and NED™. The kit also contains a tube of GeneScan™ 400 HDTM ROX™ Size Standard.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: FAM, HEX™, NED, ROX
Product Line: GeneScan™
Shipping Condition: Wet Ice
For Use With (Equipment): 3100 Genetic Analyzer

Contents & storage

Contains DNA control standard for validation of the 3100 Genetic Analyzer for fragment analysis. Store the kit at 2 - 8°C. Do not freeze.