Frames and Accessories for Immuno Breakable Modules, BreakApart frame
Frames and Accessories for Immuno Breakable Modules, BreakApart frame
Frames and Accessories for Immuno Breakable Modules, BreakApart frame
Frames and Accessories for Immuno Breakable Modules, BreakApart frame
Thermo Scientific™
Frames and Accessories for Immuno Breakable Modules, BreakApart frame
Use Thermo Scientific™ Frames and Accessories for Immuno Breakable Modules with Nunc™ C8 Breakable Modules.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeNo. per PackNo. per CasePlate Compatibility
431615Frame560Nunc C8 Breakable Modules
465404Frame560Immuno Standard Modules
Catalog number 431615
Price (USD)
413.31
Online exclusive
542.01
Save 128.70
Case of 60
Product Type:
Frame
No. per Pack:
5
No. per Case:
60
Plate Compatibility:
Nunc C8 Breakable Modules
Price (USD)
413.31
Online exclusive
542.01
Save 128.70
Case of 60
Use Thermo Scientific™ Frames and Accessories for Immuno Breakable Modules with Nunc™ C8 Breakable Modules. These frames include tools to facilitate removing the strips.
Warranty: 90 days
Specifications
DescriptionHigh optical quality for reliable results
For Use With (Application)For solid phase immuno assays
FormatStrip Plate
No. per Case60
No. per Pack5
Packaging60 Cases, 5 Pk.
Plate CompatibilityNunc C8 Breakable Modules
Product TypeFrame
Array8 x 12
ColorWhite
MaterialPolystyrene
No. of Wells96
Well ShapeCurved Edges, Flat Bottom (C-well)
Unit SizeCase of 60
Frequently asked questions (FAQs)
Which 96-well Immuno plates or modules do you recommend for various applications?
The following list offers a brief description of the features of Nunc products and their specific applications:
- Nunc-Immuno Plates, MaxiSorp surface - These plates are designed for solid phase immuno assays and have a polystyrene surface with high affinity for polar groups and hydrophilic molecules. These 96-well plates are available with flat (F), round (U), or (C) bottom-well designs.
- Nunc-Immuno Plates, PolySorp surface - These plates have a polystyrene surface which adsorps less polar molecules compared to the MaxiSorp surface and has a high affinity for hydrophobic groups. These 96-well plates are available with flat (F), round (U), or (C) bottom-well designs.
- Nunc-Immuno Modules - These modules are designed for solid-phase immuno assays. The modules are available in 8-, 12- or 16-well formats with (F), (U), and (C) bottom-well and 8-well BreakApart with (C) bottom-well designs. These different formats allow one to choose the style which is appropriate for their assay design.
BreakApart Modules and LockWell Modules can be used for radioimmunoassays.
- Nunc StarWell Immuno Breakable Modules - The 8-well modules feature eight fins on the inner wall of the (C) bottom wells. This design increases the surface area by 50%. The increase in surface area allows more molecules to be immobilized, increasing assay signal. The fin configuration provides shorter diffusion distance to the surface, reducing incubation times.
- LockWell Immuno Breakable Modules - Each plate consists of 1 x 8 breakable strips. These strips are assembled in a designed frame which locks each well into place by a spring lock. This spring lock design orientates each well at the same horizontal level allowing uniform washing and reading. The LockWell Modules are available with round (U), (C) or StarWell bottom-well designs.
- Plates and Modules with (CovaLink) Covalent Binding Surfaces: These plates can be used to bind proteins, peptides, DNA or carbohydrates. Covalent linkage occurs via secondary amine group. This allows a specific orientation and provides improved stability compared to passive adsorption due to chemical binding between molecules. Binding of molecules on PolySorp and MaxiSorp surfaces is obtained by passive adsorption. CovaLink is a surface grafted with secondary amino groups (<NH) which serves as a bridge for covalent coupling. The covalent binding can immobilize small molecules such as biotin or peptides which may otherwise bind weakly by physical adsorption and facilitate the recognition by detection molecule.
- Nunc FluoroNunc/LumiNunc 96-Well Plates: These plates are optimized for IFMA (Immunofluorometric Assays) or FIA (Fluorometric Immuno Assays). The transparent polystyrene plates give a low background fluorescence and are optimal where a read-through system is used. The white plates/modules provide maximum reflection of fluorescence signal while maintaining low background. The white plates/modules are often used for epifluorescence reading. Black modules reduce background fluorescence and minimize back-scatter light which is often encountered in epifluorescence.