Thermo Scientific™

FMAT™ 2-color Standard Microsphere Kit

Catalog number:  4316618

Related applications:

Cellular Imaging

Back to top

Description

FMAT® 2-Color Standard Microsphere Kit is designed to demonstrate the use of the FMAT® System and how to set up, run and analyze a two-color multiplexed experiment. The FMAT® 2-Color Standard Microsphere Kit contains eight vials of colored beads as follows: Dye A: Intensity 1 (highest), Intensity 2, Intensity 3 and Intensity 4 (lowest). Dye B: Intensity 1 (highest), Intensity 2, Intensity 3 and Intensity 4 (lowest). Each kit contains sufficient beads to run ten 96-well plates.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Bead Type: Standard Beads (for installation)
Product Line: FMAT™
Calibration Type: Calibration Standards
For Use With (Equipment): FMAT™ 8100 HTS System, 8200 Cellular Detection System
Product Type: Microsphere Kit
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).