MicroAmp™ Optical 96-Well Reaction Plate
Applied Biosystems™

MicroAmp™ Optical 96-Well Reaction Plate

Applied Biosystems™ MicroAmp™ Optical 96-Well Reaction Plate는 신속하고 효율적인 PCR 증폭에 최고의 온도 정확성과 일관성을 제공하는 최적의 제품입니다. 튼튼한 단편자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
4316813500 Plates
N801056010 Plates
카탈로그 번호 4316813
제품 가격(KRW)
4,599,000
온라인 행사
Ends: 31-Dec-2026
5,410,000
할인액 811,000
Each
수량:
500 Plates
제품 가격(KRW)
4,599,000
온라인 행사
Ends: 31-Dec-2026
5,410,000
할인액 811,000
Each
Applied Biosystems™ MicroAmp™ Optical 96-Well Reaction Plate는 신속하고 효율적인 PCR 증폭에 최고의 온도 정확성과 일관성을 제공하는 최적의 제품입니다. 튼튼한 단편 polypropylene을 사용해 96-well 형식으로 만들어진 이 plate는 Applied Biosystems™ 96-Well Real-Time PCR System과 Thermal Cyclers에 호환됩니다.

• cycling block의 well에서 형광 간섭을 최소화하며 동결됩니다.
• auto-fluorescent plate를 줄이도록 선별검사하였습니다.
• 고유 디자인으로 대기 온도를 차단해 well 간 온도 균일성을 보장합니다.

Oil-free PCR 증폭
MicroAmp™ 일회용 소포품은 신속한 Oil-free PCR 증폭에 있어 최고의 온도 정확성과 균일성을 제공하도록 최적화되어 있습니다. 본사 고유의 MicroAmp™ tube는 고성능 PCR 에 필수적으로 최고의 온도 및 샘플 증폭 균일성을 보장합니다.
사양
인증/적합성Certified DNA/RNase Free
용도(장비)Thermal Cycler, Genetic Analyzer, DNA Analyzer
포장500 Plates
제품 유형Microplate
수량500 Plates
반응 속도Standard
스커트 스타일Semi
부피(미터법) 웰0.3 mL
부피(미터법) 작동0.2 mL
바코드No Barcode
색상Optical
물질Polypropylene
웰 수96
제품라인MicroAmp
Unit SizeEach
구성 및 보관
Contains 50 packages of 10 plates each (each package is equivalent to catalog # N8010560)

Will require either caps or full plate cover to seal (not included)

May be stored at room temperature. With proper storage, plates may be stored indefinitely.

자주 묻는 질문(FAQ)

Are the MicroAmp Optical 96-Well Reaction Plates and MicroAmp Optical 384 -Well Reaction Plates autoclavable?

Our microplates are certified to be DNA, RNase and PCR inhibitor free as long as they are kept in the original packaging bag. They are not meant to be autoclaved.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Are the MicroAmp Optical 96-Well Reaction Plate with Barcode (Cat. No. 4306737, 4326659) and the one without Barcode (Cat. No. N8010560, 4316813) the same product?

Yes, both the plates are the same. The only difference between them is the barcode.

Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.

Can I place a standard MicroAmp 96-Well Optical Reaction Plate onto a 7500 Fast Real-Time PCR System?

No. The plate does not fit and will result in the crushing of the wells and possible contamination of the block due to leaking of the well contents.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Is the MicroAmp Optical 96-Well Reaction Plate (Cat. No. N8010560, 4316813) compatible with the SeqStudio Genetic Analyzer?

Yes, the MicroAmp Optical 96-Well Reaction Plate (Cat. No. N8010560, 4316813) is compatible with the SeqStudo Genetic Analyzer.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.

Is the 96 well plate in SequalPrep Normalization Plate Kit (Cat. No. A1051001) the same as the MicroAmp Optical 96-Well Reaction Plate (Cat. No. N8010560, 4316813)?

No, they are not the same. The MicroAmp Optical 96-Well Reaction Plate is an empty optical plate that is used for sample processing (i.e. amplification using the thermal cycler or sample loading onto a capillary instrument). The MicroAmp plate cannot be used for performing normalization as it is not coated using the ChargeSwitch technology.