4,4'-Methylenebis(cyclohexyl isocyanate), 90%, mixture of isomers, AcroSeal™
4,4'-Methylenebis(cyclohexyl isocyanate), 90%, mixture of isomers, AcroSeal™
4,4'-Methylenebis(cyclohexyl isocyanate), 90%, mixture of isomers, AcroSeal™
4,4'-Methylenebis(cyclohexyl isocyanate), 90%, mixture of isomers, AcroSeal™
Thermo Scientific Chemicals

4,4'-Methylenebis(cyclohexyl isocyanate), 90%, mixture of isomers, AcroSeal™

CAS: 5124-30-1 | C15H22N2O2 | 262.35 g/mol
数量:
100 mL
1 L
パッケージ:
AcroSeal™ Glass Bottle
製品番号(カタログ番号) 431791000
価格(JPY)
-
数量:
100 mL
パッケージ:
AcroSeal™ Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5124-30-1
IUPAC Name1-isocyanato-4-[(4-isocyanatocyclohexyl)methyl]cyclohexane
Molecular FormulaC15H22N2O2
InChI KeyKORSJDCBLAPZEQ-UHFFFAOYSA-N
SMILESO=C=NC1CCC(CC2CCC(CC2)N=C=O)CC1
さらに表示
Refractive index1.4960 to 1.4980 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=88 %
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only