Product Image
Applied Biosystems™

CBL,15FT Mac SerialW/Ferrrite

Have Questions?
Catalog NumberQuantity
43185511 Unit
Catalog number 4318551
Price (USD)
30.32
Each
Quantity:
1 Unit
Price (USD)
30.32
Each
Each package contains 15 ft of Mac Serial W⁄Ferrite cable for the 310 Genetic Analyzer.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)310 Genetic Analyzer
Product TypeMac Serial with Ferrite Cable
Quantity1 Unit
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store at room temperature.