Genomic DNA Purification Tray I

Catalog number: 4318641

Invitrogen™  Related applications: DNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This 96-well Genomic DNA (gDNA) Purification Tray is used for the isolation of both genomic DNA and total RNA from homogenized plant or animal tissue samples on the ABI PRISM® 6100 Nucleic Acid PrepSation and 6700 Automated Nucleic Acid Workstation. It is also suitable for direct DNA isolation from food and bacterial samples. The Genomic DNA (gDNA) Purification Tray is used for the TransPrep™ Chemistry protocol.

Used With
DNA Elution Solution, DNA Precipitation Solution 1, DNA Precipitation Solution 2, DNA Wash Solution 1, DNA Wash Solution 2.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Dish Type: Tray
For Use With (Equipment): 6100 Nucleic Acid PrepStation, 6700 Automated Nucleic Acid Workstation
Product Line: Applied Biosystems™
Product Size: 10 trays
Shipping Condition: Room Temperature
Type: Tray

Contents & storage

Each package contains 10 trays.

Documents

Manuals & protocols