3,5-Dimethylphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™
3,5-Dimethylphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™
3,5-Dimethylphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™
Thermo Scientific Chemicals

3,5-Dimethylphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™

CAS: 34696-73-6 | C8H9BrMg | 209.37 g/mol
Have Questions?
Catalog NumberQuantityPackaging
43194050050 mLAcroSeal™ Glass Bottle
Catalog number 431940500
Price (USD)
163.65
Special offer
Online exclusive
Ends: 28-Dec-2026
192.00
Save 28.35
Each
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
163.65
Special offer
Online exclusive
Ends: 28-Dec-2026
192.00
Save 28.35
Each
Chemical Identifiers
CAS34696-73-6
IUPAC Namebromo(3,5-dimethylphenyl)magnesium
Molecular FormulaC8H9BrMg
InChI KeyXPXQQJMQESWIKM-UHFFFAOYSA-M
SMILESCC1=CC([Mg]Br)=CC(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Yellow to brown
Titration Grignard0.40 to 0.60 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only