trans-2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)styrene, 97%
trans-2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)styrene, 97%
trans-2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)styrene, 97%
Thermo Scientific Chemicals

trans-2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)styrene, 97%

CAS: 83947-56-2 | C14H19BO2 | 230.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 432030010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS83947-56-2
IUPAC Name4,4,5,5-tetramethyl-2-[(1E)-2-phenylethenyl]-1,3,2-dioxaborolane
Molecular FormulaC14H19BO2
InChI KeyARAINKADEARZLZ-ZHACJKMWSA-N
SMILESCC1(C)OB(OC1(C)C)\C=C\C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to light yellow
Appearance (Form)Liquid or low melting solid
Infrared spectrumConforms
GC>=94.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only