Tetrabutylammonium chloride hydrate, 96%
Tetrabutylammonium chloride hydrate, 96%
Tetrabutylammonium chloride hydrate, 96%
Thermo Scientific Chemicals

Tetrabutylammonium chloride hydrate, 96%

Phase transfer catalyst | CAS: 37451-68-6 | C16H36ClN | 277.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
View all
Glass Bottle
Catalog number 432061000
Price (USD)
250.00
Each
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
250.00
Each
Chemical Identifiers
CAS37451-68-6
IUPAC Nametetrabutylazanium chloride
Molecular FormulaC16H36ClN
InChI KeyNHGXDBSUJJNIRV-UHFFFAOYSA-M
SMILES[Cl-].CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder or crystals and/or
Appearance (Form)chunks or fused mass
Infrared spectrumConforms
Titration Argentometric95 to 105 % (On dried substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only