(3,3,3-Trifluoropropyl)chlorodimethylsilane, 95%, Thermo Scientific Chemicals
(3,3,3-Trifluoropropyl)chlorodimethylsilane, 95%, Thermo Scientific Chemicals
(3,3,3-Trifluoropropyl)chlorodimethylsilane, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(3,3,3-Trifluoropropyl)chlorodimethylsilane, 95%, Thermo Scientific Chemicals

CAS: 1481-41-0 | Molecular Formula: C5H10ClF3Si | Molecular Weight: 190.67 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4322100505 gGlass bottle
Catalog number 432210050
Price (USD)
202.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
202.00
Each
Chemical Identifiers
CAS1481-41-0
IUPAC Namechlorodimethyl(3,3,3-trifluoropropyl)silane
Molecular FormulaC5H10ClF3Si
InChI KeyKBAZUXSLKGQRJF-UHFFFAOYSA-N
SMILESC[Si](C)(Cl)CCC(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to straw yellow
Infrared spectrumConforms
GC>=94 %
Appearance (Form)Liquid
Refractive index1.3720 to 1.3780 (20°C, 589 nm)

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only