Pierce™ Dextran Desalting Columns (5K MWCO)
Thermo Scientific™

Pierce™ Dextran Desalting Columns (5K MWCO)

Ready-to-use, gel-filtration Thermo Scientific Pierce Dextran Desalting Columns can separate proteins and macromolecules from low-molecular weight buffer salts and reagents up to 5000 Daltons (5 kDa) in 5- or 10-mL volumes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
4323310 mL
432305 mL
Catalog number 43233
Price (USD)
184.00
Each
Volume (Metric):
10 mL
Request bulk or custom format
Price (USD)
184.00
Each
Thermo Scientific Pierce Dextran Desalting Columns are ready-to-use, disposable, gel-filtration columns for separating proteins and other macromolecules from low molecular-weight buffer salts and reagents.

Applications
• Removing salts from protein solutions
• Eliminating phenol from nucleic acid preparations
• Separating excess crosslinker from conjugate preparations
• Removing excess derivatizing agents from modified proteins
• Withdrawing unreacted dye from fluorescent antibodies
• Eliminating free radiolabel from labeled proteins
• Buffer exchange

Gravity-flow gel filtration using dextran desalting columns
Gravity-flow gel filtration involves the chromatographic separation of molecules of different dimensions based on their relative abilities to penetrate a suitable stationary phase. A chromatographic matrix, usually consisting of very small, uncharged porous particles in an aqueous solution, is packed into a column and then used for separation via size exclusion. Different levels of separation can be achieved based on the pore size of the medium packed into the desalting column—in our case, dextran. With size exclusion filtration, large molecules are excluded from the internal pores of the gel and emerge from the column first. Smaller molecules can penetrate the pores, then progress through the column at a slower rate. These smaller molecules are subsequently flushed through the column with additional buffer volume. Our Dextran Desalting Columns are rigid for easy handling and provide excellent flow properties. Dextran is stable in water, salt solutions, organic solvents and alkaline or weakly acidic solutions. Dextran is also heat-stable and can be autoclaved dry or in solution at a neutral pH for 30 minutes at 120°C without affecting its chromatographic properties.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
DisposableSingle-use
Product TypeDesalting Column
Purification TargetBuffer Exchange, Protein
Quantity5 Columns
Volume (Metric)10 mL
Column TypeSize-exclusion, Dextran Resin
FormatGravity Flow Column
MWCO5.0 kDa
Product LinePierce
Unit SizeEach
Contents & Storage
Store at room temperature or 4°C.

Frequently asked questions (FAQs)

Besides precipitation with ammonium sulfate, what are some ways I can concentrate my protein sample?

You may remove excess solvent and smaller moieties by centrifugation through a microconcentrator. This will concentrate your protein sample.

(1) Choose microconcentrator tube (available from a variety of commercial suppliers) with a protein cutoff smaller than the molecular weight of the protein in the sample. Check our Pierce Protein Concentrators PES.

(2) Add 1 µL of 20% w/v SDS to a dry microconcentrator tube (if sample does not already contain SDS).

(3) Slowly add sample (a few microliters at a time) to membrane until membrane is completely wet. Centrifuge to near (but not complete) dryness.

(4) If intention is to desalt sample or buffer exchange: Add ~50 µL water to microconcentrator and spin until nearly dry. Repeat buffer exchange. Sample will remain on membrane. Check our Zeba desalting proteins.

(5) Using a new collection tube, invert membrane and spin at low speed (1000 x g) to elute protein from membrane. Add 2X SDS-Sample Buffer containing 10 mM DTT to membrane: vortex, invert and spin. Final volume should be ~20 µL.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.