2-Methylsulfanylpyrimidine-4-carboxylic acid, 97%
2-Methylsulfanylpyrimidine-4-carboxylic acid, 97%
2-Methylsulfanylpyrimidine-4-carboxylic acid, 97%
Thermo Scientific Chemicals

2-Methylsulfanylpyrimidine-4-carboxylic acid, 97%

CAS: 1126-44-9 | C6H6N2O2S | 170.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 432370050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1126-44-9
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Appearance (Color)White to light yellow
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only