• Find Real-Time and Digital PCR technical tips, troubleshooting help, and resources within our new Real-Time PCR and Digital PCR Applications Support Center.
TaqMan™ Universal PCR Master Mix, no AmpErase™ UNG
Applied Biosystems™

TaqMan™ Universal PCR Master Mix, no AmpErase™ UNG

TaqMan Universal PCR Master Mix, No AmpErase UNG, is the ideal reagent solution when you need a master mix forRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
43240181 x 5 mL, 200 reactions
432402010 x 5 mL, 2,000 reactions
43266141 x 50 mL 2,000 reactions
43643412 x 5 mL, 400 reactions
43643435 x 5 mL, 1,000 reactions
Catalog number 4324018
Price (USD)
685.65
Online exclusive
754.00
Save 68.35
Each
Quantity:
1 x 5 mL, 200 reactions
Price (USD)
685.65
Online exclusive
754.00
Save 68.35
Each
TaqMan Universal PCR Master Mix, No AmpErase UNG, is the ideal reagent solution when you need a master mix for multiple 5' nuclease DNA applications. Applied Biosystems reagents have been validated with TaqMan assays and Applied Biosystems real-time systems to ensure sensitive, accurate, and reliable performance every time.

• Contains AmpliTaq Gold DNA Polymerase to provide a better yield and a more robust 5' nuclease assay than AmpliTaq DNA Polymerase
• Includes a passive internal reference based on proprietary ROX dye, for superb precision on Applied Biosystems real-time PCR instruments
• Buffer enhancements guarantee performance and reliability in all applications, even with G/C-rich sequences
• Contains dUTP in the dNTP mix to protect against carry-over contamination, when used with AmpErase UNG
• Rapid assay development guidelines are provided to minimize optimization time

Alternative product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve taken the best of TaqMan Universal PCR Master Mix and added additional capabilities for your gene expression analysis.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)7000 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, GeneAmp 9600, StepOne™, Standard Mode, StepOnePlus™, Standard Mode
No. of Reactions200 Reactions
Passive Reference DyeROX (Pre-mixed)
PolymeraseAmpliTaq Gold DNA Polymerase
Product LineTaqMan
Product TypeUniversal PCR Master Mix
Quantity1 x 5 mL, 200 reactions
Sample TypeDNA (Genomic), cDNA
Shipping ConditionWet Ice
Sufficient For200 Reactions
Volume5 mL
Detection MethodPrimer-probe
For Use With (Application)Gene Expression, Genotyping, miRNA analysis
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Supplied at 2X concentration. The mix contains AmpliTaq Gold™ DNA Polymerase, dNTPs with dUTP, Passive Reference 1 and optimized buffer components. This pack contains one 5 mL tube, sufficient for 200 reactions at the 50 μL total volume. Store at 2-8°C

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

Frequently asked questions (FAQs)

What master mix can I use with the TaqMan SNP Genotyping Assays?

We recommend using either the TaqPath ProAmp Master Mix or the TaqMan Genotyping Master Mix. The TaqMan Genotyping Master Mix has the advantage of proven performance with up to 3 days of pre- and post-PCR stability, allowing you to set up plates ahead of time or read the plates later (see the data here, https://tools.thermofisher.com/content/sfs/brochures/cms_039236.pdf) while the TaqPath ProAmp Master Mix can handle samples that may have inhibitors present.

What does it mean when a miRNA target entry has been removed (obsoleted) from miRBase?

miRBase continues to re-annotate the miRNA sequences as more information becomes available. A sequence that once mapped to a given species (e.g.: human) may no longer exist, so it is removed from miRBase. Accordingly, the original assay for that sequence no longer maps to an annotated miRNA, and as a result the annotation fields on the search results page on the Thermo Fisher Scientific portal are empty. An example of this is Assay “hsa-miR-505” (Assay ID 001049). miRBase continues to re-annotate the miRNA sequences as more information becomes available. A sequence that once mapped to a given species (e.g.: human) may no longer exist, so it is removed from miRBase. Accordingly, the original assay for that sequence no longer maps to an annotated miRNA, and as a result the annotation fields on the search results page on the Thermo Fisher Scientific portal are empty. An example of this is Assay “hsa-miR-505” (Assay ID 001049).

What is the TaqMan MicroRNA Assay?

A TaqMan MicroRNA Assay has two components: an RT reaction containing a miRNA specific stem-loop reverse-transcription primer and a separate specific TaqMan miRNA Assay. The RT stem-loop primer provides the specificity for the mature miRNA target; it does not detect its precursor. The formation of a RT primer/mature miRNA-chimera extends the length of the 5’ end of the miRNA. The longer RT product provides a miRNA specific cDNA template amenable to the TaqMan assay design.

What is mechanism of the TaqMan MicroRNA Assays?

There are 2 steps involved in using the TaqMan MicroRNA Assays:

1. RT step: total RNA to cDNA using miRNA-specific RT primers from the TaqMan MicroRNA Assays and the TaqMan MicroRNA Reverse Transcription Kit (P/N 4366596, 200 reactions or 4366597, 1000 reactions)

2. PCR step: cDNA to PCR products using primers and probe from the TaqMan MicroRNA Assays together with TaqMan Universal PCR Master Mix No AmpErase UNG (P/N 4324018).

Why are miRNAs so important?

MicroRNAs are believed to play an important role in post transcriptional gene regulation. A total of ~850 unique miRNAs have been discovered so far, including ~333 human miRNAs. They are relatively abundant, ranging from a few to as many as 50,000 molecules per cell. They account for approximately 1% of the predicted genes in animals and plants. The level of individual miRNAs varies dramatically across cell type and developmental stages. These differences in miRNA level are believed to be a key indicator of miRNA activity.

Recent research has implicated miRNAs in the following: cell development, differentiation, communication and cell death; DNA methylation and chromatin modification; metabolism; human cancer development; haematopoiesis; nervous system patterning; insulin secretion.