1-Bromo-4-(1,1-difluoroethyl)benzene, 95%
1-Bromo-4-(1,1-difluoroethyl)benzene, 95%
1-Bromo-4-(1,1-difluoroethyl)benzene, 95%
Thermo Scientific Chemicals

1-Bromo-4-(1,1-difluoroethyl)benzene, 95%

CAS: 1000994-95-5 | C8H7BrF2 | 221.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
Packaging:
Glass bottle
Catalog number 432530050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1000994-95-5
IUPAC Name1-bromo-4-(1,1-difluoroethyl)benzene
Molecular FormulaC8H7BrF2
InChI KeyQXIBKCFAFRHORF-UHFFFAOYSA-N
SMILESCC(F)(F)C1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=94.0 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only