Trans-4-(Trifluoromethyl)cyclohexanecarboxylic acid, 98%
Trans-4-(Trifluoromethyl)cyclohexanecarboxylic acid, 98%
Trans-4-(Trifluoromethyl)cyclohexanecarboxylic acid, 98%
Thermo Scientific Chemicals

Trans-4-(Trifluoromethyl)cyclohexanecarboxylic acid, 98%

CAS: 133261-33-3 | C8H11F3O2 | 196.17 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4326000505 gGlass bottle
Catalog number 432600050
Price (USD)
714.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
714.00
Each
Chemical Identifiers
CAS133261-33-3
IUPAC Name4-(trifluoromethyl)cyclohexane-1-carboxylic acid
Molecular FormulaC8H11F3O2
InChI KeyLMEAZIIFLVDISW-UHFFFAOYSA-N
SMILESOC(=O)C1CCC(CC1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point152°C to 157°C
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only