Cyclopropylzinc bromide, 0.5M solution in THF, AcroSeal™
Cyclopropylzinc bromide, 0.5M solution in THF, AcroSeal™
Cyclopropylzinc bromide, 0.5M solution in THF, AcroSeal™
Thermo Scientific Chemicals

Cyclopropylzinc bromide, 0.5M solution in THF, AcroSeal™

CAS: 126403-68-7 | C3H5BrZn | 186.37 g/mol
Have Questions?
Catalog NumberQuantityPackaging
43282050050 mLAcroSeal™ Glass Bottle
Catalog number 432820500
Price (USD)
280.65
Online exclusive
312.00
Save 31.35
Each
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
280.65
Online exclusive
312.00
Save 31.35
Each
Chemical Identifiers
CAS126403-68-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Light brown to black
Titration Grignard0.30 to 0.60 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only