FMAT Blue™ Monofunctional Reactive Dye Labeling Kit

Catalog number:  4328853

Applied Biosystems™  Related applications: Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The FMAT Blue® Monofunctional Dye Labeling Kit is supplied as a NHS-ester. The dye is a channel 1 dye emitting in the 650 nm range. The dye is designed to label antibodies, proteins and other biopolymers that have a primary amine, for use as secondary detection reagents.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 8200 Cellular Detection System, FMAT® 8100 HTS System
Label or Dye: FMAT Blue®
Product Line: FMAT Blue®
Product Size: 1 kit

Contents & storage

The FMAT Blue® labeling kit contains the dye and desalting columns. Dye is light sensitive.
Store unopened vials at -20 ° in the dark. Store desalting columns at room temperature.

Documents

Manuals & protocols