Applied Biosystems™

ABI PRISM™ 7000 Sequence Detection Systems Spectral Calibration Kit

Catalog number: 4328895
Applied Biosystems™

ABI PRISM™ 7000 Sequence Detection Systems Spectral Calibration Kit

Catalog number: 4328895
Catalog Number
4328895
Unit Size
8 x 96 well plates
Price (USD)
Price: 1,454.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
43288958 x 96 well plates
Price: 1,454.00
Your Price:
-

Recurring order eligible

Save time by having your items shipped automatically.

Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This Applied Biosystems™ easy-to-use kit establishes the pure dye spectra and multicomponent values for FAM™, JOE™, NED™, ROX™, SYBR™ Green, VIC™, and TAMRA™ dyes on the ABI PRISM™ 7000 Sequence Detection System. The ABI PRISM™ 7000 Sequence Detection Systems Spectral Calibration Kit includes eight optical 96-well reaction plates: one preloaded and sealed background plate, and seven preloaded and sealed spectral calibration plates with seven separate dye standards.
• 96-well optical reaction plates preloaded with dyes (FAM™, JOE™, NED™, ROX™, SYBR™ Green, VIC™, and TAMRA™) are provided for quick and easy calibration of the ABI PRISM™ 7000 Sequence Detection System
• Easy-to-use and provides accurate discrimination of the fluorescence signal of individual dyes in the reaction mixture
• Includes the Passive Reference I dye standard for signal normalization in all real-time and endpoint reactions Spend Your Time on Research—Not Instrument Calibration This kit's easy-to-use, preloaded, 96-well optical reaction plates enable users to calibrate the ABI PRISM™ 7000 Sequence Detection System quickly and easily. Insert the reaction plates during system installation, and the system automatically collects the dye standard's spectral information. The system's application algorithm then uses the calibration information to ensure accurate data analysis.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Calibrated Dye
FAM, JOE, NED, ROX, TAMRA™, TMR (Tetramethylrhodamine), VIC, SYBR™ Green
For Use With (Equipment)
7000 System
Detection Method
Fluorescent
Product Line
ABI PRISM™
Product Type
Spectral Calibration Kit
For Use With (Application)
Real Time PCR (qPCR)
Shipping Condition
Dry Ice
Calibration Type
Spectral Calibration

Contents & Storage

Contains one Background plate, and seven Spectral Calibration plates with seven dye standards (FAM™, JOE™, NED™,ROX™, SYBR™ Green, VIC™, TAMRA™). Store at -15 to 25°C. Protect from light.