7900HT Fast Real-Time PCR System with 384-Well Block Module
This product was discontinued in 2012. We recommend QuantStudio qPCR product family. Learn more
7900HT Fast Real-Time PCR System with 384-Well Block Module
Applied Biosystems™

7900HT Fast Real-Time PCR System with 384-Well Block Module

The 7900HT Fast Real-Time PCR System is a real-time quantitative PCR system that combines 96- and 384-well plate compatibility andRead more
Have Questions?
Catalog NumberQuantity
43290011 instrument
Catalog number 4329001
Price (USD)
-
Request A Quote
Quantity:
1 instrument
The 7900HT Fast Real-Time PCR System is a real-time quantitative PCR system that combines 96- and 384-well plate compatibility and the TaqMan™ Low Density Array with fully automated robotic loading-and now also offers optional Fast real-time PCR capability.

• Fast PCR option reduces run time to about 35 minutes in a standard 96-well format, or about 55 minutes in a 384-well plate
• Continuous wavelength detection from 500-660 nm allows the use of multiple fluorophores in a single reaction
• Measure gene expression levels, detect and quantitate pathogens, perform allelic discrimination (SNP genotyping) assays as well as score the presence of gene sequences
• 96- or 384-well plate compatibility (including the TaqMan™ Low Density Array)
• Optional Enterprise edition software provides data analysis tools that support 21 CFR Part 11 guidelines providing data integrity and security
• Hands-free plate-loading and unloading provides true walkaway automation allowing you to increase your lab's productivity
• Proven assay development guidelines save time and money

High Throughput
The 7900HT System is a high-throughput real-time PCR system that detects and quantitates nucleic acid sequences. An optional Automation Accessory (not included) combined with 384-well plate capability make the 7900HT system ideally suited to meet the high-throughput requirements of today's drug discovery process. Key applications include gene expression quantitation and the detection of single nucleotide polymorphisms (SNPs) using the fluorogenic 5' nuclease assay.

Flexibility
The 7900HT system is a versatile research tool that can accommodate any real-time PCR need. User-interchangeable thermal cycling block formats let you select the format that’s right for your project, using industry-standard 96- and 384-well formats, as well as a novel 384-well TaqMan™ Low Density Array and a new Fast 96-well block that reduces run times from 2 hours to about 30 minutes. An easy automation upgrade path lets you add features to meet throughput demands. A Fast PCR option reduces run times from 2 hours to about 30 minutes.

Powerful Software
Using two automation software tools, Plate Utility and Automation Controller, most of the gene expression and SNP genotyping workflow is fully automated for high-throughput applications and requires minimal user intervention. You can now process 5,000 x 384 well plates (384 markers), which equates to 1.92 million genotypes. Using Relative Quantification (RQ) Manager, you can also process 200 x 384-well plates (96 detectors with quadruplicate data points⁄multiplexed endogenous control), which equates to 153,600 data points. Fully integrated into one complete Enterprise system, SNP Manager and RQ Manager eliminate data analysis bottlenecks in high-throughput SNP and gene expression research by allowing significantly more plates to be analyzed simultaneously. This reduces hands-on analysis time and simplifies the data analysis workflow.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Calibrated DyeSYBR™ Green I, ROX™, NED™, JOE™, TAMRA™, VIC™, FAM™, TET™
Detection MethodSYBR, Primer-Probe Detection
Dimensions72 cm (W) x 84 cm (D) x 64 cm (H) (with Drawer Open)
Dynamic RangeUp to 9 Logs of Linear Dynamic Range
For Use With (Equipment)7900HT Fast System, AB 7900 HT 384-Well System
Format384-well Plate
High-throughput CompatibilityHigh-throughput Compatible, Multiplexing
OpticsExtended-life 488 nm Argon-ion Laser Excitation Source, No Filters (CCD acts as spectrograph with continuous detection 500 to 650 nm)
Passive Reference DyeROX (Separate Tube), No ROX, Other Passive Reference Dye, ROX (Pre-mixed)
Quantity1 instrument
Reaction SpeedFast
Reaction Volume Range5–20 μL
Run Time52 min. (384-well in Fast Mode), <2 hrs (384-well in Standard Mode)
Sample Ramp RateFast Mode: ± 2.2°C⁄s, Standard mode: ± 1.6°C⁄s
SensitivityDown to 10 Copies
Temperature Range (Metric)4 to 100°C
Thermal Accuracy±0.25°C of Display Temperature
Thermal Cycling SystemPeltier-based System
Thermal Uniformity±0.5°C
Throughput≥5,000 Real Time PCR rxns per day (384-well block and automation accessory), ≥10,000 endpoint PCR rxns per hour (384-well block and automation accessory)
Weight114 kg with Automation Accessory, 82 kg without Automation Accessory
Block FormatInterchangeable
CapacityUp to 84 x 384-well Plates with Optional Automation Accessory
Unit SizeEach
Contents & Storage
1- Instrument with 384-well Block Module

Frequently asked questions (FAQs)

What is the threshold on the Applied Biosystems real-time PCR instruments and how do I set it?

The threshold is the numerical value assigned for each run that reflects the average dRn (change in fluorescence) during the initial cycles of PCR (baseline). The threshold is set by determining a statistically significant point (or value) above the baseline. You can manually adjust the threshold should you desire, to the place in the geometric phase where your replicates are tightest. For more information on how to manually set a threshold, please refer to the tutorial entitled Data Analysis on the ABI PRISM 7700: Setting Baselines and Thresholds (P/N 4370923). While this tutorial is based on the ABI PRISM 7700 system, the concepts are still valid for the Applied Biosystems real-time PCR instruments. You can search the document on our website by using the part number above as the keyword.

If you choose not to manually adjust the baselines and thresholds, the SDS software on the Applied Biosystems real-time PCR instruments has an Auto Ct algorithm that can be used to automatically generate baseline and threshold values for individual detectors. The algorithm calculates baseline and threshold parameters for a detector based on the assumption that the data exhibits the "typical" amplification curve. Experimental error (i.e. contamination, pipetting inaccuracies) can produce amplification curves that deviate significantly from a typical amplification curve. The data from these irregularities can affect the Auto Ct algorithm by causing it to generate incorrect baseline and threshold parameters for the associated detector. Therefore, Applied Biosystems recommends that after analysis of experimental data, you review all baseline and threshold parameters determined by the Auto Ct algorithm.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Is the Applied Biosystems 7900HT SDS Automation Controller software and Zymark Twister Instrument compatible with MicroAmp Fast Optical 96-Well Reaction Plates with Barcode (P/N 4346906)?

No. The current version of the Zymark Twister Instrument and SDS Automation Controller software is NOT compatible with MicroAmp Fast 96-Well Optical Plates with Barcode. The Zymark Twister Instrument is compatible with MicroAmp Optical 96-Well Reaction Plate with Barcode, MicroAmp Optical 384-Well Reaction Plate with Barcode, as well as TaqMan Low Density Arrays.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Why is my Applied Biosystems 7900HT SDS run file “read only” protected after the run has completed when using the SDS Automation Controller software and the Zymark Twister Instrument?

The file remains "read only" due to the status of the SDS Automation Controller Queue. The SDS run file in question still remains in the queue and must be cleared by the user within the SDS Automation Controller Software. To do this, the customer must open the SDS Automation Controller software, highlight the plate in question, select File -> Clear Selected Plates.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

What is the default user name and password for logging into the SDS 2.3 software?

When launching the SDS 2.3 software, you will be prompted for a user name and password. The default user name will be administrator and the default password field can be left blank. The user can simply click "OK" to launch the SDS 2.3 software. User accounts are configured by selecting TOOLS and then clicking LOCAL USER ACCOUNT MANAGER. Additional information regarding the LOCAL USER ACCOUNT MANAGER can be found by searching SDS 2.3 Online Help from the HELP pull down menu in the SDS 2.3 software.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Where can I find a protocol to perform Fast TaqMan Real-Time PCR using the 384-well block on the Applied Biosystems 7900HT?

For detailed instructions on performing Fast TaqMan Real-Time PCR using the 384-well block on Applied Biosystems 7900HT, please refer to User Bulletin for Performing Fast TaqMan Gene Quantitation using 384 well plates on the Applied Biosytems 7900HT Fast-Real-Time PCR System with the 384 well Block Module. (P/N 4369584)

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.