4-Methoxybenzenesulfonamide, 95%
4-Methoxybenzenesulfonamide, 95%
4-Methoxybenzenesulfonamide, 95%
Thermo Scientific Chemicals

4-Methoxybenzenesulfonamide, 95%

CAS: 1129-26-6 | C7H9NO3S | 187.213 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 432910250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1129-26-6
IUPAC Name4-methoxybenzene-1-sulfonamide
Molecular FormulaC7H9NO3S
InChI KeyMSFQEZBRFPAFEX-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)S(N)(=O)=O
さらに表示
Appearance (Color)White to off-white
Infrared spectrumConforms
HPLC>=94.0 %
Appearance (Form)Crystalline powder
Melting point109°C to 113°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only