2-Metoxibenzonitrilo, 99 %, Thermo Scientific Chemicals
2-Metoxibenzonitrilo, 99 %, Thermo Scientific Chemicals
2-Metoxibenzonitrilo, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Metoxibenzonitrilo, 99 %, Thermo Scientific Chemicals

CAS: 6609-56-9 | C8H7NO | 133.15 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 g
25 g
Envase:
Frasco de vidrio
Número de catálogo 433160050
Precio (USD)
-
Cantidad:
5 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS6609-56-9
IUPAC Name2-methoxybenzonitrile
Molecular FormulaC8H7NO
InChI KeyFSTPMFASNVISBU-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1C#N
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid or low melting solid
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only