Dextran sulfate sodium salt, approx. M.W. 500,000
Dextran sulfate sodium salt, approx. M.W. 500,000
Dextran sulfate sodium salt, approx. M.W. 500,000
Thermo Scientific Chemicals

Dextran sulfate sodium salt, approx. M.W. 500,000

CAS: 9011-18-1 ∣ (C6H7O5)m(C6H7O5)n ∣ 40,000 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 433241000
Price (USD)
272.65
Special offer
Online exclusive
Ends: 28-Dec-2026
321.00
Save 48.35
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
272.65
Special offer
Online exclusive
Ends: 28-Dec-2026
321.00
Save 48.35
Each
Chemical Identifiers
CAS9011-18-1
IUPAC NameDextran sulfate sodium salt
MDL NumberMFCD00081551
SpecificationsSpecification SheetSpecification Sheet
Sulfated ash40 % to 50 %
Appearance (Form)Powder
Solubilitycolorless to light yellow
pH5 to 7.5 (1 % in water)
Appearance (Color)Off-white to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only