Proteinase K from the fungus Engyodontium album is a non-specific serine protease that is useful for general digestion of proteins.ProteinaseRead more
Have Questions?
Catalog Number
Quantity
4333793
5 mL
Catalog number 4333793
Price (USD)
166.65
Online exclusive
181.00
Save 14.35
Each
Quantity:
5 mL
Price (USD)
166.65
Online exclusive
181.00
Save 14.35
Each
Proteinase K from the fungus Engyodontium album is a non-specific serine protease that is useful for general digestion of proteins.
Proteinase K Remains Active: • Over a wide pH range (optimal activity between 6.5 and 9.5) • Under denaturing conditions (e.g. the presence of SDS or urea) • In the presence of metal chelating agents (e.g. EDTA) • At comparatively high temperatures (optimum digestion temperature is 65°C)
Applications: Removal of endogenous nucleases during the preparation of DNA and RNA; preparation of tissue sections for in situ hybridization.
Performance and Quality Testing: Endodeoxyribonuclease and exodeoxyribonuclease assays; liquid form tested for absence of RNase activity.
Unit Definition: One mAnson unit is described as that amount of enzyme that liberates 1 μmole of Folin-positive amino acid within one minute at 37°C using hemoglobin as a substrate.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Chromatin Biology/RNA Extraction/DNA Extraction
Product TypeProteinase K
PurityDNase-Free, RNase-Free
Quantity5 mL
Shipping ConditionWet Ice
FormLiquid
Unit SizeEach
Contents & Storage
Store in refrigerator (2°C to 8°C).
Frequently asked questions (FAQs)
How do you recommend storing proteinase K?
We recommend storing it at 4°C.
How do I isolate DNA from single bacterial colonies before PCR?
Scrape a single colony from the surface of an agar plate and transfer to 12 µL of SCL solution [10 mM Tris-HCl (pH 7.5), 1 mM EDTA, 50 µg/mL proteinase K] and incubate for 15 min at 55 degrees C. Inactivate proteinase K for 15 min at 80 degrees C and add 20 µL of deionized water. Centrifuge at 12,000 x g for 3 min. Transfer the supernatant to a new tube.