Product Image
Applied Biosystems™

Autosampler Installation Kit (Septa), 384-well plate

Have Questions?
Catalog NumberQuantity
43361341 Kit
Catalog number 4336134
Price (USD)
-
Quantity:
1 Kit
The 384-Well Plate Autosampler Kit (for use with septa) for 3730 and 3730xl DNA Analyzer. This kit includes the plate base, retainer, septa and 384-well microplates. This system enables plates to be attached to the instrument autosampler.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Sequencing
For Use With (Equipment)3730 DNA Analyzer
Product TypeAutosampler Installation Kit
Quantity1 Kit
Format384-well Plate
Unit SizeEach
Contents & Storage
This kit includes the plate base, retainer, septa and 384-well microplates.