Lithium diphenylphosphide, 0.5M solution in THF, AcroSeal™
Lithium diphenylphosphide, 0.5M solution in THF, AcroSeal™
Lithium diphenylphosphide, 0.5M solution in THF, AcroSeal™
Lithium diphenylphosphide, 0.5M solution in THF, AcroSeal™
Thermo Scientific Chemicals

Lithium diphenylphosphide, 0.5M solution in THF, AcroSeal™

CAS: 65567-06-8 | C12H10LiP | 192.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
433621000100 mLAcroSeal™ Glass Bottle
Catalog number 433621000
Price (USD)
223.65
Online Exclusive
248.00
Save 24.35 (10%)
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
223.65
Online Exclusive
248.00
Save 24.35 (10%)
Each
Chemical Identifiers
CAS65567-06-8
IUPAC Namelithiodiphenylphosphane
Molecular FormulaC12H10LiP
InChI KeyWKUYEGHEUWHKIU-UHFFFAOYSA-N
SMILES[Li]P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration excess HCl0.40 to 0.60 M
Appearance (Form)Liquid
Appearance (Color)Orange to deep red
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only