BigDye™ Terminator v1.1 Cycle Sequencing Kit
Applied Biosystems™

BigDye™ Terminator v1.1 Cycle Sequencing Kit

This BigDye™ Terminator v1.1 Cycle Sequencing Kit is designed for specialty applications that require optimal basecalling adjacent to the primer and for sequencing short PCR product templates with rapid electrophoresis run modules.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
43374525000 Reactions
433744924 Reactions
4337450100 Reactions
43374511000 Reactions
Catalog number 4337452
Price (USD)
-
Quantity:
5000 Reactions

This BigDye™ Terminator v1.1 Cycle Sequencing Kit is designed for specialty applications that require optimal basecalling adjacent to the primer and for sequencing short PCR product templates with rapid electrophoresis run modules.

  • Improve the quality of your results for a wide range of sequencing applications
  • Sequence challenging templates and sequences more successfully
  • Get longer, higher-quality reads with more uniform peak heights and optimal signal balance
  • Enhance your productivity and reduce costs

Tackle Tough Sequences
With the BigDye Terminator v1.1 Cycle Sequencing Kit you get a robust, highly flexible chemistry for a wide range of applications, including de novo sequencing and resequencing.

Get More Uniform Peak Heights, Improved Accuracy
With BigDye Terminator v1.1 kit's superior chemistry, you generate data with uniform peak heights and optimized signal balance-which results in longer, higher-quality reads. You also get more accurate base assignments for heterozygote and mutation detection.

Easily Integrate into Your Workflow
BigDye™ Terminator v.1.1 kit's chemistry requires no new software or instrument recalibration.You can easily integrate either kit into your current workflow with minimal changes to your protocols.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Application)Standard Sequencing
For Use With (Equipment)SeqStudio™ Genetic Analyzer, SeqStudio™ Flex Series Genetic Analyzer, 3500/3500xL Genetic Analyzer, 3730/3730xl DNA Analyzer Thermal Cyclers: GeneAmp™ PCR System 9700 Dual Series, Veriti™ Fast 96‐Well and 384‐Well Thermal Cyclers
Product LineBigDye™ Terminator
Product TypeSequencing Buffer
Quantity5000 Reactions
TechniqueFluorescent Dye Terminator Sequencing
Template CompatibilityBAC DNA, Plasmid DNA (≤15 Kb), PCR Amplicons, Single Stranded DNA
Format12-well Strips, 8-well Strip, 96- or 384-well Plate
Unit SizeEach
Contents & Storage
Each kit contains:
• 2 x 20 mL tubes of BigDye™ Terminator v1.1 Ready Reaction Mix
• 2 tubes of M13 (-21) Primer
• 2 tubes of pREF-BDT™ Control DNA (200 ng/μL)
• 1 x 28 mL tube of 5X Sequencing Buffer (may be stored at 4°C).

Store kit at -15°C to -25°C and avoid repeated freeze-thaw cycles.

Frequently asked questions (FAQs)

What information should I include when submitting data to Genetic Analysis Technical Support?

When contacting Genetic Analysis Support (FAS, FSE, Technical Support), please have (if possible):

Instrument model and serial numbers
Version of the software are you using
Application and/or kit information
Kit lot number
Number of runs on the capillary or array
Polymer type and lot number
Buffer lot number
Hi-Di Formamide lot number or loading solution information
Sample data that can be sent to the support person
Details on:
  -Reaction setup (i.e., how much DNA was used, primer, etc.)
  -How the reactions were cleaned up (alcohol precipitation, columns, etc)
  -What template DNA was used (i.e. plasmid, PCR product)
  -How the template DNA was prepared?

All of this information will help the support person quickly and accurately assess the problem and provide you with recommendations for resolution.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

I have a long homopolymer stretch in my sample that I cannot get through. Do you have any tips for getting through it?

In order to get through homopolymeric regions, you can either use anchored sequencing primers or try using the dRhodamine Terminator Cycle Sequencing Kit since it uses ddTTP instead of ddUTP and has been shown to be less prone to producing stutters, specifically with poly-T regions.

For more information on sequencing through homopolymer stretches, please refer to the DNA Sequencing by Capillary Electrophoresis: Applied Biosystems Chemistry Guide: Second Edition (Cat. No. 4305080, Rev. C). The guide can be found by searching the Thermo Fisher Scientific website with the catalog number 4305080.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

I am trying to sequence GC-rich DNA without success. Do you have any tips for getting through this?

When trying to sequence through GC-rich regions, the following tips may improve your success:

• Set the reaction up at 1X or 0.5X strength. Heavily diluting the BigDye Terminator Ready Reaction mix can reduce the success of the sequecing reaction through long GC stretches.

• Perform a hot start at 98–99°C for 5 minutes.

• Use 5% (w/v) DMSO or freshly made betaine in the reaction.

• Use the dGTP BigDye Terminator Cycle Sequencing Kit. G peaks will appear compressed due to the presence of ddGTP, but sequencing through long GC stretches using the dGTP kit is typically more successful than with the standard BigDye Terminator Cycle Sequencing Kits (containing ddITP). For more information on sequencing GC-rich DNA, please refer to the DNA Sequencing by Capillary Electrophoresis: Applied Biosystems Chemistry Guide: Second Edition (Cat. No. 4305080, Rev. C). The guide can be found by searching the Thermo Fisher Scientific website with the catalog number 4305080.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

I see another sequence running under mine. What can cause this?

Some of the causes of another sequence appearing under the primary sequence are:

• Contamination: There is more than one species of DNA present (e.g., multiple PCR products).

• Primers: Primer is annealing in more than one location on the template, primer dimer, primer degradation or not manufactured properly resulting in N+1 or N-1, carry over from PCR reaction, or primers pairs in a multiplex reaction that are not appropriate for multiplexing (i.e., primers anneal inappropriately).

• Spectral/Matrix: If the raw data signal intensity of the sample is too high or saturated it can exceed the amount of color bleedthrough (or spectral overlap) that the matrix (310) or spectral (3130, 3730, 3500) are removing, resulting in secondary peaks appearing in a very specific pattern (e.g., a red peak always appearing under a green peak). A change in the camera, laser, or optical alignment requires that a new matrix be made or a new spectral calibration be performed.

For more information on more than one sequence or set of peaks in a sequencing run, please refer to the DNA Sequencing by Capillary Electrophoresis: Applied Biosystems Chemistry Guide: Second Edition (Cat. No. 4305080, Rev. C). The guide can be found by searching the Thermo Fisher Scientific website with the catalog number 4305080.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

My data seems top heavy. It starts out strong and then gets weaker. What can cause this?

The cause of this is usually an overabundance of template DNA relative to the amount of BigDye Terminator Ready Reaction mix being used in the reaction. This can drive the reaction to incorporate the labeled ddNTPs closer to the 5’ end of the sequence. To balance the signal, either increase the amount of BigDye Terminator Ready Reaction mix being used in the reaction or decrease the concentration of the template DNA.

For more information on other causes of short reads and how to address each issue, please refer to the DNA Sequencing by Capillary Electrophoresis: Applied Biosystems Chemistry Guide: Second Edition (Cat. No. 4305080, Rev. C). The guide can be found by searching the Thermo Fisher Scientific website with the catalog number 4305080.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.