Heat Seal Film for Sequencing & Fragment Analysis Sample Plates

Catalog number: 4337570

Invitrogen™  Related applications: Sanger Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This film is used to seal sample plates during sequencing and fragment analysis.

Used With
MicroAmp® Optical 384-Well Reaction Plate with Barcode, MicroAmp® Optical 384-Well Reaction Plate with Barcode, MicroAmp® Optical 96-Well Reaction Plate, MicroAmp® Optical 96-Well Reaction Plate with Barcode.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color: Clear
Plate Compatibility: 384-Well Plates, 96-Well Plates
Product Size: 1 package
Shipping Condition: Room Temperature
Type: Film, Seal, or Applicator

Contents & storage

Each package contains enough film for approximately 3,500 seals.

Documents

Manuals & protocols